missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Erbium(III) nitrate pentahydrate, 99.9% (trace metal basis), ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 315140250

 View more versions of this product

Catalog No. AC315140250


 

Specifications Description & Specifications

Specifications

Crystals or Crystalline Powder
443.35
Pink
Er(NO3)3·5H2O
Erbium(III) nitrate pentahydrate
MFCD00149691
Plastic bottle
Trace Metal Basis
25g
33 to 39% (Er)
99.9%
ErN3O9·5H2O
0.1% max.
10031-51-3

Chemical Identifiers

10031-51-3
ErN3O9·5H2O
443.35
Erbium(III) nitrate pentahydrate
Trace Metal Basis
99.9%
Er(NO3)3·5H2O
MFCD00149691
33 to 39% (Er)

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger