missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Erbium(III) nitrate hydrate, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1292022

 View more versions of this product

Catalog No. AA1292022



Specifications

Specifications

Erbium(III) nitrate hydrate
(REO)
MFCD00010988
acmc-20al2v
LWHHUEHWVBVASY-UHFFFAOYSA-N
erbium(3+);trinitrate;pentahydrate
53249207
Crystalline
Hygroscopic
100g
10031-51-3
ErH10N3O14
UN1477
Typically insoluble in water.
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Er+3]
443.346
353.27 (Anhydrous)
99.9%
130°C (in -4H2O)

Chemical Identifiers

10031-51-3
443.346
LWHHUEHWVBVASY-UHFFFAOYSA-N
53249207
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Er+3]
ErH10N3O14
MFCD00010988
acmc-20al2v
erbium(3+);trinitrate;pentahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

DOTInformation : Hazard Class: 5.1; Packaging Group: II

EINECSNumber : 233-436-7

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents