Please login to your online account to display your discounted pricing

Alfa Aesar™ Erbium(III) iodide, anhydrous, 90% min, (REO)

Chemical Identifiers

Chemical Name or Material Erbium(III) iodide
CAS 13813-42-8
Name Note Anhydrous
Formula Weight 547.97
MDL Number MFCD00016075
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3354004

  SDS
alfa aesar™
3354004
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3354009

  SDS
alfa aesar™
3354009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 547.97
Assay Percent Notes (REO)
MDL Number MFCD00016075
TSCA TSCA
CAS 13813-42-8
Melting Point 1020°C
Boiling Point 1280°C
Chemical Name or Material Erbium(III) iodide
Name Note Anhydrous
Assay ≤90%
Sensitivity Hygroscopic
Quantity 2g
Physical Form Crystalline