Please login to your online account to display your discounted pricing

Alfa Aesar™ Erbium(III) chloride, ultra dry, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Erbium(III) chloride
CAS 10138-41-7
Name Note Ultra dry
Formula Weight 273.62
MDL Number MFCD00010989
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3573914

  SDS
alfa aesar™
3573914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3573906

  SDS
alfa aesar™
3573906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 273.62
Packaging Ampule
Chemical Name or Material Erbium(III) chloride
Storage Note 1
  • Ampouled under argon
MDL Number MFCD00010989
Health Hazard 1 Warning
Odor Odorless
Physical Form Powder
Name Note Ultra dry
CAS 10138-41-7
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Hygroscopic
Quantity 25g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
TSCA TSCA
RTECS Number KD8575000
Boiling Point 1500°C
Assay Percent Notes (REO)
Assay 99.9%
Melting Point 774°C

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

  • Ampouled under argon