missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Erbium(III) chloride, anhydrous, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 8991730

 View more versions of this product

Catalog No. AA8991730



Specifications

Specifications

Erbium (III) chloride
10138-41-7
(metals basis)
MFCD00010989
Cl[Er](Cl)Cl
273.609
273.62
Odorless
1500°C
250g
Anhydrous
99.9%
Cl3Er
HDGGAKOVUDZYES-UHFFFAOYSA-K
trichloroerbium
66277
Beads
Hygroscopic
774°C

Chemical Identifiers

10138-41-7
273.609
HDGGAKOVUDZYES-UHFFFAOYSA-K
trichloroerbium
Cl3Er
MFCD00010989
66277
Cl[Er](Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : KD8575000

TSCA : TSCA

SDS Documents