Please login to your online account to display your discounted pricing

Alfa Aesar™ Erbium(III) carbonate hydrate, 99.99%

$40.69 - $115.12

Chemical Identifiers

Chemical Name or Material Erbium(III) carbonate hydrate
CAS 22992-83-2
Molecular Formula Er2(CO3)2·xH2O
Formula Weight 514.54 (Anhydrous)
MDL Number MFCD00149951
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1720903

  SDS
alfa aesar™
A1720903
1g Each for $40.69

AAA1720906

  SDS
alfa aesar™
A1720906
5g Each for $115.12
Description & Specifications

Specifications

Formula Weight 514.54 (Anhydrous)
MDL Number MFCD00149951
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material Erbium(III) carbonate hydrate
TSCA TSCA
Assay 99.99%
Molecular Formula Er2(CO3)2·xH2O
Health Hazard 1 Warning
CAS 22992-83-2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.