Please login to your online account to display your discounted pricing

Alfa Aesar™ Erbium(III) bromide nonahydrate, 99.99%

$129.92 - $526.43

Chemical Identifiers

Chemical Name or Material Erbium(III) bromide nonahydrate
CAS 29843-93-4
Molecular Formula ErBr3·9H2O
Formula Weight 569.11 (406.97 Anhydrous)
MDL Number MFCD00209649
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1803506

 
alfa aesar™
A1803506
5g Each for $129.92

AAA1803514

 
alfa aesar™
A1803514
25g Each for $526.43
Description & Specifications

Specifications

Formula Weight 569.11 (406.97 Anhydrous)
Chemical Name or Material Erbium(III) bromide nonahydrate
TSCA TSCA
Assay 99.99%
Molecular Formula ErBr3·9H2O
MDL Number MFCD00209649
CAS 29843-93-4
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
It is used, like other metal bromide compounds, in water treatment, chemical analysis and for certain crystal growth applications.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.