Please login to your online account to display your discounted pricing

Alfa Aesar™ Erbium(III) acetate tetrahydrate, REacton™, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Erbium(III) acetate tetrahydrate
CAS 15280-57-6
Purity Grade Reacton™
Linear Formula Er(OOCCH3)3·4H2O
Formula Weight 416.45 (344.39 Anhydrous)
MDL Number MFCD00013034
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4048106

 
alfa aesar™
4048106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4048114

 
alfa aesar™
4048114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4048122

 
alfa aesar™
4048122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 416.45 (344.39 Anhydrous)
Chemical Name or Material Erbium(III) acetate tetrahydrate
TSCA TSCA
Odor Odorless
Purity Grade Reacton™
Physical Form Crystalline Aggregates
Assay 99.9%
Sensitivity Hygroscopic
Assay Percent Notes (REO)
Linear Formula Er(OOCCH3)3·4H2O
MDL Number MFCD00013034
CAS 15280-57-6
Quantity 5g
Density 2.114g/mL