missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Erbium(III) acetate tetrahydrate, 99% (REO)

Manufacturer:  Alfa Aesar™ 4589722

 View more versions of this product

Catalog No. AA4589722



Specifications

Specifications

Erbium (III) acetate tetrahydrate
99%
C6H9ErO6·4H2O
MFCD00013034
Crystalline Aggregates
Odorless
Pink
15280-57-6
(REO)
Er(OOCCH3)3·4H2O
416.45 (344.39 Anhydrous)
2.114g/mL
Hygroscopic
100g

Chemical Identifiers

15280-57-6
C6H9ErO6·4H2O
MFCD00013034
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents