missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Erbium foil, 0.25mm (0.01 in.) thick, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 12386FF

 View more versions of this product

Catalog No. AA12386FF



Specifications

Specifications

Erbium foil
25 x 25mm
7440-52-0
(REO)
MFCD00010987
UYAHIZSMUZPPFV-UHFFFAOYSA-N
erbium
23980
Foil
≈1.4g/25x25mm
0.25mm (0.01 in.) thick
Odorless
Er
unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic
[Er]
167.259
CHEBI:33379
99.9%

Chemical Identifiers

7440-52-0
167.259
UYAHIZSMUZPPFV-UHFFFAOYSA-N
23980
erbium
Er
MFCD00010987
unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic
CHEBI:33379
[Er]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents