missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Erbium foil, 0.025mm (0.001 in.) thick, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 12388FF

 View more versions of this product

Catalog No. AA12388FF



Description

Description

  • Packaged under argon
Specifications

Specifications

Erbium foil
25 x 25 mm
7440-52-0
Er
UN3178
Insoluble in water.
[Er]
167.259
CHEBI:33379
99.9%
≈0.14g/25x25mm
0.025mm (0.001 in.) thick
(REO)
MFCD00010987
chips
UYAHIZSMUZPPFV-UHFFFAOYSA-N
erbium
23980
Foil

Chemical Identifiers

7440-52-0
167.259
UYAHIZSMUZPPFV-UHFFFAOYSA-N
23980
erbium
Er
MFCD00010987
chips
CHEBI:33379
[Er]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

EINECSNumber : 231-160-1

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

Documents