Please login to your online account to display your discounted pricing

erbB4 Mouse anti-Human, Mouse, Biotin, Clone: HFR1, eBioscience™

Mouse Monoclonal Antibody

$220.40 - $368.30

Specifications

Antigen erbB4
Clone HFR1
Host Species Mouse
Gene Alias c-erbB4
Species Reactivity Human, Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122589

 
life technologies
LS13968780
25μg Each for $220.40

501122590

 
life technologies
LS13968782
100μg Each for $368.30
Description & Specifications

Specifications

Antigen erbB4
Clone HFR1
Host Species Mouse
Gene Alias c-erbB4
Species Reactivity Human, Mouse
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody HFR1 recognizes human and mouse epidermal growth factor receptor 4 (ErbB4 or HER4). ErbB4 belongs to the family of receptor tyrosine kinases (RTK) of which HER1 (EGRF), HER2 (ErbB2), and HER3 (ErbB3) are members. RTKs function to affect a variety of cell processes including proliferation, differentiation, migration, survival and apoptosis and are often disregulated in cancer. The ErbB4 protein contains extracellular ligand binding domains, a transmembrane region, and a cytoplasmic tail containing the tyrosine kinase domain and the binding site for the HFR1 antibody. Upon ligand binding, ErbB4 homo- or hetero-dimerizes with other family members, becomes phosphorylated, and can be cleaved allowing translocation to the nucleus. The HRF1 antibody recognizes membrane-bound, cytoplasmic and nuclear ErbB4. Cytosolic ErbB4 expression was found to correlate with a positive prognosis in a subset of breast cancer patients, while nuclear ErbB4 expression was inversely correlated with tumor grade and aggressiveness of breast tumors.