Invitrogen™ ErbB4 (HER4) DFS Item
Oncogene ERBB4 (HER4) is a member of the type I receptor protein tyrosine subfamily that includes EGFR, ERBB2, and ERBB3.
Manufacturer:  Invitrogen™ LSPV4104
 View more versions of this product
Catalog No. PV4104
Description
Description
ERBB4 binds and is activated by neuregulins and other factors and induces a variety of cellular responses including mitogenesis and differentiation.
Specifications
Specifications
NP_005226
Frozen
61.6kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
Kinase Assay
EGF Receptor (ErbB) Kinase Family
GST Tag
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
ErbB4 (HER4)
Baculovirus
Human
Documents