Please login to your online account to display your discounted pricing

ER 27319 maleate, Tocris

Selective Syk kinase inhibitor

$177.37 - $734.99

Chemical Identifiers

Chemical Name or Material ER 27319 maleate
Assay Percent Range >99%
Molecular Formula C18H20N2O·C4H4O4
Formula Weight 396.17
Synonym 10-(3-Aminopropyl)-3, 4-dimethyl-9(10H)-acridinone maleate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

247110

 
r & d systems
247110
10mg Each for $177.37

247150

 
r & d systems
247150
50mg Each for $734.99
Description & Specifications

Specifications

Formula Weight 396.17
Quantity 10mg
Assay Percent Range >99%
CAS 1204480-26-1
Chemical Name or Material ER 27319 maleate
Molecular Formula C18H20N2O·C4H4O4
Synonym 10-(3-Aminopropyl)-3, 4-dimethyl-9(10H)-acridinone maleate
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C