missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ EPPS, 0.2M buffer soln., pH 7.5

$64.24 - $110.85

Chemical Identifiers

CAS 16052-06-5
Molecular Formula C9H20N2O4S
Molecular Weight (g/mol) 252.329
MDL Number MFCD00006160
InChI Key OWXMKDGYPWMGEB-UHFFFAOYSA-N
Synonym HEPPS
PubChem CID 85255
ChEBI CHEBI:42298
IUPAC Name 3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
SMILES C1CN(CCN1CCCS(=O)(=O)O)CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60511AE View Documents Alfa Aesar™
J60511AE
100mL Each for $64.24
 
AAJ60511AK View Documents Alfa Aesar™
J60511AK
250mL Each for $110.85
 
Specifications

Chemical Identifiers

16052-06-5
252.329
OWXMKDGYPWMGEB-UHFFFAOYSA-N
85255
3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
C9H20N2O4S
MFCD00006160
HEPPS
CHEBI:42298
C1CN(CCN1CCCS(=O)(=O)O)CCO

Specifications

16052-06-5
MFCD00006160
HEPPS
C1CN(CCN1CCCS(=O)(=O)O)CCO
252.329
CHEBI:42298
7.5
EPPS buffer soln.
C9H20N2O4S
0.2M buffer soln.
OWXMKDGYPWMGEB-UHFFFAOYSA-N
3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
85255
Liquid
100mL
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Keep cold