missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ EPPS, 0.2M buffer soln., pH 7.5

Manufacturer:  Alfa Aesar™ J60511AK

 View more versions of this product

Catalog No. AAJ60511AK



Specifications

Specifications

EPPS buffer soln.
C9H20N2O4S
0.2M buffer soln.
OWXMKDGYPWMGEB-UHFFFAOYSA-N
3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
85255
Liquid
250mL
16052-06-5
MFCD00006160
HEPPS
C1CN(CCN1CCCS(=O)(=O)O)CCO
252.329
CHEBI:42298
7.5

Chemical Identifiers

16052-06-5
252.329
OWXMKDGYPWMGEB-UHFFFAOYSA-N
85255
3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
C9H20N2O4S
MFCD00006160
HEPPS
CHEBI:42298
C1CN(CCN1CCCS(=O)(=O)O)CCO
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Keep cold

Documents