missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ EPPS, 0.2M buffer soln., pH 7.0

$82.35 - $128.92

Chemical Identifiers

CAS 16052-06-5
Molecular Formula C9H20N2O4S
Molecular Weight (g/mol) 252.329
MDL Number MFCD00006160
InChI Key OWXMKDGYPWMGEB-UHFFFAOYSA-N
Synonym HEPPS
PubChem CID 85255
ChEBI CHEBI:42298
IUPAC Name 3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
SMILES C1CN(CCN1CCCS(=O)(=O)O)CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61538AE View Documents Alfa Aesar™
J61538AE
100mL Each for $82.35
 
AAJ61538AK View Documents Alfa Aesar™
J61538AK
250mL Each for $128.92
 
Description

Description

Applications
EPPS is a commonly used Good’s buffer. It is often used as separator in ultrathin isoelectric focusing gels and enhances the resolution of phosphoglucomutase. It is used as a buffering agent in biology and biochemistry.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

16052-06-5
252.329
OWXMKDGYPWMGEB-UHFFFAOYSA-N
85255
3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
C9H20N2O4S
MFCD00006160
HEPPS
CHEBI:42298
C1CN(CCN1CCCS(=O)(=O)O)CCO

Specifications

16052-06-5
MFCD00006160
HEPPS
C1CN(CCN1CCCS(=O)(=O)O)CCO
252.329
CHEBI:42298
7.0
EPPS buffer soln.
C9H20N2O4S
0.2M buffer soln.
OWXMKDGYPWMGEB-UHFFFAOYSA-N
3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
85255
Liquid
100mL
Safety and Handling

Safety and Handling

TSCA : TSCA