Eppendorf™ Varispenser™ plus/Varispenser Bottle-Top Reagent Dispensers
Bottle-top reagent dispenser for taking aliquots of liquid from large supply bottles
$1,038.73 - $2,174.68
Specifications
Autoclavable Yes
Certifications/Compliance EN ISO 8655
Product Type Bottletop Reagent Dispenser
Precision ≤0.1%
Warranty 2 Years
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Accuracy Dispensing Interval Includes Volume Range Price Quantity  
Catalog Number Mfr. No. Accuracy Dispensing Interval Includes Volume Range Price Quantity  
13-687-263 Eppendorf™
4960000043
±0.5% 0.5mL Telescopic tube, 3 adapters for 32, 38, 40mm O.D. bottle thread 5 to 25mL Each for $1,383.89
13-687-266 Eppendorf™
4961000012
±0.6% 0.05mL Telescopic tube, 3 adapters for 28, 40, 45mm O.D. bottle thread, recirculation valve, valve toggle and media specific fine adjustment 0.5 to 2.5mL Each for $1,097.07
13-687-270 Eppendorf™
4961000055
±0.5% 1mL Telescopic tube, 3 adapters for 32, 38, 40mm O.D. bottle thread, recirculation valve, valve toggle and media specific fine adjustment 10 to 50mL Each for $1,511.91
13-687-260 Eppendorf™
4960000019
±0.6% 0.05mL Telescopic tube, 3 adapters for 28, 40, 45mm O.D. bottle thread 0.5 to 2.5mL Each for $1,038.73
13-687-261 Eppendorf™
4960000027
±0.5% 0.1mL Telescopic tube, 3 adapters for 28, 40, 45mm O.D. bottle thread 1 to 5mL Each for $1,038.73
13-687-264 Eppendorf™
4960000051
±0.5% 1mL Telescopic tube, 3 adapters for 32, 38, 40mm O.D. bottle thread 10 to 50mL Each for $1,451.93
13-687-268 Accessories Eppendorf™
4961000039
±0.5% 0.2mL Telescopic tube, 3 adapters for 28, 40, 45mm O.D. bottle thread, recirculation valve, valve toggle and media specific fine adjustment 2 to 10mL Each for $1,097.07
13-687-265 Eppendorf™
4960000060
±0.5% 2mL Telescopic tube, 3 adapters for 32, 38, 40mm O.D. bottle thread 20 to 100mL Each for $2,116.34
13-687-262 Accessories Eppendorf™
4960000035
±0.5% 0.2mL Telescopic tube, 3 adapters for 28, 40, 45mm O.D. bottle thread 2 to 10mL Each for $1,038.73
13-687-269 Eppendorf™
4961000047
±0.5% 0.5mL Telescopic tube, 3 adapters for 32, 38, 40mm O.D. bottle thread, recirculation valve, valve toggle and media specific fine adjustment 5 to 25mL Each for $1,443.84
13-687-271 Eppendorf™
4961000063
±0.5% 2mL Telescopic tube, 3 adapters for 32, 38, 40mm O.D. bottle thread, recirculation valve, valve toggle and media specific fine adjustment 20 to 100mL Each for $2,174.68
13-687-267 Accessories Eppendorf™
4961000020
±0.5% 0.1mL Telescopic tube, 3 adapters for 28, 40, 45mm O.D. bottle thread, recirculation valve, valve toggle and media specific fine adjustment 1 to 5mL Each for $1,097.07
Specifications
Description
  • PFA-sealing of the slide piston prevents jamming Fine adjustment depending on viscosity of liquid No crystallization of liquid
  • No dripping when valve closed Telescopic tube for use with high bottles Rapid volume setting using precise graduation scale Patented safety valve for deairing without loss of reagent Cannel-sealing cap prevents media contact Autoclavable
  • Extremely chemical-resistant
  • Varispenser plus features safety valve and fine adjustment
Order Info
Bottle-top dispenser fits on external threads of 32mm (sizes 1, 2 and 3) or 45mm (sizes 4, 5 and 6), complete with telescopic tube, wrench and 3 adapters.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Yes
Bottletop Reagent Dispenser
2 Years
EN ISO 8655
≤0.1%