missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ Vacufuge™ Concentrator

Three aeration modes to concentrate DNA/RNA, proteins, and other samples quickly

Manufacturer:  Eppendorf™ 022820001

 View more versions of this product

Catalog No. 07-748-13


Alert:

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.


Description

Description

Combines centrifugal force and vacuum. For gentle concentration of DNA, RNA, nucleotides, proteins, and samples for gel electrophoresis.

  • Available as a basic instrument for use with existing pumps (with minimum vacuum pressure of 20mbar) or as a complete system with a maintenance-free PTFE vacuum pump (40L/min.) and vapor trap
  • Three aeration modes (aqueous, alcohol, and off) maximize evaporation efficiency, prolong pump life span, and reduce processing time by up to 20%
  • Corrosion-resistant stainless-steel chamber withstands chemical exposure
  • Maximum speed/force: 1400rpm/240xG
  • Brake function brings rotor to a faster stop
  • Space-saving design with low noise level of ≤60dBA for the compact laboratory
  • Choice of four heating levels (ambient, 30°, 45° and 60°C) provides optimal evaporation speed for your samples

Order Info

Concentrator is supplied with a stackable 48-place fixed-angle rotor for 1.5–2.0mL microcentrifuge tubes. Additional rotors are available separately. Nonstackable rotors for 96-well plates, 15mL conical tubes, and vials are also available.

Specifications

Specifications

30, 45, 60°C
120V
9 in.
12.625 in.
115V 50/60Hz
9 in.
37cm
240 x G
37.47 lb.
17kg
12.62 in.
48 x 1.5 to 2.0mL tubes
48 x 1.5 to 2mL Fixed angle rotor (increasing capacity up to 144 samples), transparent lid
50/60Hz
14.5 in.
Basic Unit Model
14.5 in.
23cm
Stainless Steel
1400rpm
150w
32cm
14.5 x 12.62 x 9 in. (37 x 32 x 23cm)
SDS
Documents