Please login to your online account to display your discounted pricing

Eppendorf™ twin.tec™ Real-Time 96-Well PCR Platesc


Fluorescence intensity up to ten times higher than with frosted wells

Manufacturer: eppendorf™  951022067

 View more versions of this product

Catalog No. E951022067

  • / Case of 25 Each
 

Description & Specifications

Specifications

Autoclavable Yes
Color Black
For Use With (Application) RT-PCR applications
Material Polypropylene
Rimmed Yes
Skirt Style Semi
Sterile Yes
Well Color White

The limiting factor in low volume real-time PCR is often the remaining intensity of fluorescence. The white wells of the Eppendorf twin.tec real-time PCR plates reflect fluorescence much better than clear or frosted wells. Thus, lower levels of fluorescence are still detectable with the same instrument–just by using the right consumables. Additionally, white wells significantly reduce interfering background fluorescence and lead to increased homogeneity of replicates and reproducible results.

  • White wells for better reflection
  • High mechanical stability
  • Raised rims for effective sealing
  • Optimal heat transfer due to reduced wall thickness
  • Autoclavable (121°C, 20 min.)
  • Rigid polycarbonate frame ensures easy and reliable handling, manually or automated
  • Polypropylene wells guarantee excellent and fast temperature transfer to the sample