Please login to your online account to display your discounted pricing

Eppendorf™ twin.tec™ Real-Time 96-Well PCR Platesc


Fluorescence intensity up to ten times higher than with frosted wells

Manufacturer: eppendorf™  951022027

 View more versions of this product

Catalog No. E951022027

  • / Case of 25 Each
 

Description & Specifications

Specifications

Autoclavable Yes
For Use With (Application) RT-PCR applications
Material Polypropylene
Rimmed Yes
Skirt Style Skirted
Sterile Yes
Well Color White
Color Black
Product Type 96-Well PCR Plate
Applications RT-PCR applications
Sterile Yes
Material Polypropylene