missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ twin.tec™ 384-Well Microbiology PCR Plates

Continuous skirt for compatibility with automated systems

$413.42 - $413.42

Specifications

Format Standard
Array 16 x 24
For Use With (Equipment) Automated systems
Skirt Style Skirted
Material Polypropylene
View More Specs

Products 2
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
36-102-3837
View Documents Promotion Details
Eppendorf™
0030129342
Clear
Pack of 10 Each for $413.42
 
36-102-3838
View Documents Promotion Details
Eppendorf™
0030129350
Blue
Pack of 10 Each for $413.42
 
Description

Description

  • Skirted for bar code
  • Stackable
  • Within SBS footprint recommendations
  • Ideal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler™ Pro 384
  • 45 to 40μL max. well volume

Specifications

Specifications

Standard
Automated systems
Polypropylene
384
10/Pk.
16 x 24
Skirted
40 to 45 μL
U
SDS
missing translation for 'documents'

missing translation for 'documents'