Eppendorf™ Tube Adapters for 5415/5417 Series Microcentrifuges
$53.08 - $61.03
Specifications
For Use With (Equipment) MiniSpin™ and MiniSpin™ Plus microcentrifuge
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Bore Size (Metric) Tube Size Price Quantity  
Catalog Number Mfr. No. Bore Size (Metric) Tube Size Price Quantity  
05-400-125 Eppendorf™
022636243
0.6cm 0.25 to 0.4 mL Pack of 6 Each for $61.03
05-400-126 Eppendorf™
022636260
0.6cm .2mL PCR Pack of 6 Each for $53.08
05-400-124 Eppendorf™
022636227
0.8cm 0.5 mL Pack of 6 Each for $53.08
Specifications
Description
•    For use with fixed-angle rotors 05-406-13 and 05-406-14 and swinging bucket rotor 05-406-41 (for Model 5417R), and fixed-angle rotors 05-400-122 and 13-688-198 (for Models 5415D/5415R)
•    Adapters fit any Eppendorf™ rotor with slots to hold 1.5 to 2.0 microcentrifuge tubes with maximum 11mm O.D.
Order Info
Supplied in packs of six
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
MiniSpin™ and MiniSpin™ Plus microcentrifuge