Eppendorf™ Tube Adapters for 5415/5417 Series Microcentrifuges
For use with 5910 R/5920 R rotors
Supplier: Eppendorf™ 022636260
Description
- For use with fixed-angle rotors 05-406-13 and 05-406-14 and swinging bucket rotor 05-406-41 (for Model 5417R), and fixed-angle rotors 05-400-122 and 13-688-198 (for Models 5415D/5415R)
- Adapters fit any Eppendorf™ rotor with slots to hold 1.5 to 2.0 mL microcentrifuge tubes with maximum 11 mm O.D.
Order Info
Supplied in packs of six
Specifications
PCR | |
MiniSpin™ and MiniSpin™ Plus microcentrifuge | |
0.6 cm | |
0.2 mL |