missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eppendorf™ Top Buret Bottle-Top Burette
Extremely robust with the display and control elements guaranteeing simple, fatigue-free operation
Supplier: Eppendorf™ 4965000017
Description
Extremely robust with the display and control elements guaranteeing simple, fatigue-free operation
The innovative Top Buret from Eppendorf sets new standards for manual titration. A new, pulse-free dispensing technique allows the desired volumes to be dispensed continuously, thus eliminating the need for new liquid aspiration each time. This makes titration simpler and safer, with precision values well within required limits
- Continuous, pulse-free dispensing technique for rapid titration
- Patented safety valve for deairing without loss of reagent
- Voltage supply with long-life primary cells and charging display
- Dispensing range from 0.01mL to 999.9mL
- Removable electronics unit
- Single channel with simple calibration program
- Discharge tube rotatable by 360°
- Adjustable discharge tube: horizontal: 142 to 220mm, vertical: 10 to 200mm
- Telescopic aspiration tube for automatically setting the length from 210mm to 370mm
- Connection for dry tube
- The medium is dispensed by simply rotating the wheels
Model M
- Dispensing rate of 2,500μL per rotation
- Dispensing rate of 5,000μL per rotation
Specifications
Rapid Titration | |
25 mL | |
Recirculation valve, valve toggle, telescopic filling tube, adjustable discharge tube, three adapters 40/38/32mm bottle threads, 2 x 1.5V microbattery, quality certificate |
Yes | |
Rapid titration |