Eppendorf™ Thermomixer™ R Accessory, Thermoblock Dry Blocks
Ensure precise “fit” for common lab tubes
Manufacturer:  Eppendorf™ 022670565
 View more versions of this product
Catalog No. 05-400-207
Description
Description
These exchangeable thermoblocks for the Thermomixer R and ThermoStat provide excellent flexibility. The range of potential applications is significantly expanded due to the support for slides and microarrays, in addition to a broad range of tube sizes.
  • Shielded thermoblocks are contact-safe
  • Efficient exchange simplifies operation
  • Tubes are visible during operation and easily accessible
Specifications
Specifications
For one PCR and deepwell plate
4.9 in.
124mm
1400rpm
For one PCR and deepwell plate
Dry Block for Thermomixer R
Lid for improved temperature accuracy
70°C
Documents