Eppendorf™ Thermomixer™ R Accessory, Thermoblock Dry Blocks
Ensure precise “fit” for common lab tubes
$1,238.38 - $1,260.11
Catalog Number | Mfr. No. | Holds | For Use With (Equipment) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Holds | For Use With (Equipment) | Price | Quantity | ||||
05-400-202
|
Eppendorf™
022670506 |
24 x 0.5mL | Micro Test Tubes |
Each for $1,238.38
|
|
05-400-203
|
Eppendorf™
022670522 |
24 x 1.5mL | Micro Test Tubes |
Each for $1,238.38
|
|
0540213
|
Eppendorf™
022670514* |
4 x 50mL | Falcon™ Conical Tubes |
Each for $1,260.11
|
|
Description
These exchangeable thermoblocks for the Thermomixer R and ThermoStat provide excellent flexibility. The range of potential applications is significantly expanded due to the support for slides and microarrays, in addition to a broad range of tube sizes.
- Shielded thermoblocks are contact-safe
- Efficient exchange simplifies operation
- Tubes are visible during operation and easily accessible
Specifications
Dry Block for Thermomixer R |