missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ Stackable PCR Racks

For handling and storing tubes and plates

Supplier:  Eppendorf™ 0030124545

Catalog No. E0030124545


Explore available promotions

Description

Description

  • Compatible for 0.1, 0.2 and 0.5mL tubes and 96-well plates
  • Stackable

Specifications

Specifications

Assorted
Microtubes (0.1, 0.2, and 0.5 mL)
10/Pk.
SDS
Documents

Documents

Promotions

Promotions