Eppendorf™ Rotors for Centrifuge 5427 R
Aerosol-tight with Eppendorf QuickLock™
Manufacturer:  Eppendorf™ 5409712004
 View more versions of this product
Catalog No. 05-100-163
Description
Description
  • Max. speed: 18,210 × g (12,700 rpm)
  • Max. capacity: 48 × 1.5/2.0 mL tubes
Specifications
Specifications
48 x 1.5/2mL vessels
12,700 rpm
F-45-48-11
5427R Centrifuge Rotor
Eppendorf QuickLock™ 5427 R
Documents