Please login to your online account to display your discounted pricing

Eppendorf™ Research plus™ Adjustable-Volume Pipetters Trade-In


Resistant to chemical, thermal and mechanical stress

Manufacturer: eppendorf™  022478967

 View more versions of this product

Catalog No. TI13690032

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Resistant to chemical, thermal and mechanical stress
Color Blue
Volume (Metric) 100 to 1000μL
Accuracy ±3%, ±1%, ±0.6%
Increments 100, 500, 1000μL
Color Blue