Please login to your online account to display your discounted pricing

Eppendorf™ Research plus™ Adjustable-Volume Pipetters Trade-In


Resistant to chemical, thermal and mechanical stress

Manufacturer: eppendorf™  022478941

 View more versions of this product

Catalog No. TI13690030

  • / Each
 

Description & Specifications

Specifications

Accuracy ±2.5%, ±1%, ±0.6%
For Use With (Application) Resistant to chemical, thermal and mechanical stress
Increments 20, 100, 200μL
Color Yellow
Volume (Metric) 20 to 200μL
Color Yellow