Please login to your online account to display your discounted pricing

Eppendorf™ Research plus™ Adjustable-Volume Pipetters Trade-In


Resistant to chemical, thermal and mechanical stress

Manufacturer: eppendorf™  022478908

 View more versions of this product

Catalog No. TI13690027

  • / Each
 

Description & Specifications

Specifications

Color Yellow
Accuracy ±5%, ±1.2%, ±1%
Increments 2, 10, 20μL
For Use With (Application) Resistant to chemical, thermal and mechanical stress
Volume (Metric) 2 to 20μL