Please login to your online account to display your discounted pricing
Eppendorf™ Repeater™ stream and Xstream Pipetter
Combine easy handling with a broad range of features
Manufacturer: eppendorf™  022460811
 View more versions of this product
Catalog No. 05-401-041
  • / Each
Description & Specifications
Specifications
Battery Type Lithium Ion
Compatible Tips Eppendorf™ Combitips™ Plus tips
Number of Channels 1
Volume (Metric) Range 1μL to 50mL
Model Repeater Xstream™
Product Type With charging adapter
Volume (Metric) 1μL to 50mL
Finger Style Ergonomic
For Use With (Equipment) Eppendorf™ Combitips™ Plus tips
Includes Battery
Warranty 2 Years
Repeater stream is suitable for all standard pipetting and dispensing applications. For added versatility, the Repeater Xstream can also perform titrations, sequential dispensing and aspiration of supernatants. When used with Combitips™ Plus tips, both systems form an unbeatable and essential lab tool for all applications.
Applications
  • Electronic dispensing with programmable memory functions
  • Contamination-free dispensing using the positive displacement principle
  • Ideal for viscous solutions (e.g., glycerol, oil) or liquids with high vapor pressure (e.g., acetone, ethanol)
  • Dispensing in a long series
Features, Repeater Stream and Xstream
  • Positive displacement dispensing and pipetting
  • Automatic dispensing
  • Rapid and ergonomic dispensing with motorized piston
  • Single-button tip ejector
  • Li-ion battery
  • Function selecton wheel
  • Automatic Combitips Plus tip recognition
  • IvD conformity
  • Charging possible during operation
Additional Features of the Repeater Xstream
  • Aspirating of supernatants
  • Titration
  • Sequential dispensing
Accessories
Accessories for Catalog Number: 05401041
Item Mfr. No. Item Description Price Per Unit Quantity  
  • Get a Case of Tips When You Buy a Repeating Pipette
    Buy Now
    Expires 31-12-2016