Please login to your online account to display your discounted pricing
Eppendorf™ Repeater™ stream and Xstream Pipetter
Combine easy handling with a broad range of features
Manufacturer: eppendorf™  022460803
 View more versions of this product
Catalog No. 05-401-040
  • / Each
Description & Specifications
Specifications
Product Type With charging adapter
Model Repeater stream™
Number of Channels 1
Number of Channels 1
Battery Type Lithium Ion
Compatible Tips Eppendorf™ Combitips™ Plus tips
Finger Style Ergonomic
For Use With (Equipment) Eppendorf™ Combitips™ Plus tips
Includes Battery
Warranty 2 Years
Volume (Metric) Range 1μL to 50mL
Volume (Metric) 1μL to 50mL
Product Type Pipet
Capacity Range 1μL to 50mL
Compatibility Eppendorf™ Combitips™ Plus tips
Accessories
Accessories for Catalog Number: 05401040
Item Mfr. No. Item Description Price Per Unit Quantity