Please login to your online account to display your discounted pricing
Eppendorf™ Repeater™ stream and Xstream Pipetter
Combine easy handling with a broad range of features
$1,533.80 - $1,881.80
Specifications
Battery Type Lithium Ion
Volume (Metric) Range 1μL to 50mL
Volume (Metric) 1μL to 50mL
Finger Style Ergonomic
Includes Battery
View More Specs
Products
Catalog Number Mfr. No. Model Product Type Price Quantity    
05-401-040
  Accessories
eppendorf™
022460803
Repeater stream™ With charging adapter Each for $1,533.80
05-401-041
  Accessories
eppendorf™
022460811
Repeater Xstream™ With charging adapter Each for $1,843.14
13-683-505
eppendorf™
4986000823
 Repeater stream™ With charging adapter and bundle with charger stand Each for $1,572.46
13-683-506
eppendorf™
4986000831
Repeater Xstream™ With charging adapter and bundle with charger stand Each for $1,881.80
Description & Specifications
Specifications
Battery Type Lithium Ion
Volume (Metric) Range 1μL to 50mL
Volume (Metric) 1μL to 50mL
Finger Style Ergonomic
Includes Battery
Number of Channels 1
Warranty 2 Years
Repeater stream is suitable for all standard pipetting and dispensing applications. For added versatility, the Repeater Xstream can also perform titrations, sequential dispensing and aspiration of supernatants. When used with Combitips™ Plus tips, both systems form an unbeatable and essential lab tool for all applications.
Applications
  • Electronic dispensing with programmable memory functions
  • Contamination-free dispensing using the positive displacement principle
  • Ideal for viscous solutions (e.g., glycerol, oil) or liquids with high vapor pressure (e.g., acetone, ethanol)
  • Dispensing in a long series
Features, Repeater Stream and Xstream
  • Positive displacement dispensing and pipetting
  • Automatic dispensing
  • Rapid and ergonomic dispensing with motorized piston
  • Single-button tip ejector
  • Li-ion battery
  • Function selecton wheel
  • Automatic Combitips Plus tip recognition
  • IvD conformity
  • Charging possible during operation
Additional Features of the Repeater Xstream
  • Aspirating of supernatants
  • Titration
  • Sequential dispensing
Uses Eppendorf Combitips Plus Tips.
Two years
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.