missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ PCR Cooler

Maintains sample temperature for over an hour; changes color when temperature exceeds 7°C

Manufacturer:  Eppendorf™ 022510509

 View more versions of this product

Catalog No. 05-403-01



Description

Description

  • Maintains sample temperature at 0°C
  • Temperature of each sample is indicated by the color of its individual well
  • Cone-shaped contours prevent samples from freezing
  • Accommodates 0.2mL PCR tubes (individual, tube strip, or 96-well plate)

Specifications

Specifications

Set of 1 Pink and 1 Blue
Set of 1 Pink and 1 Blue
0.2mL
Documents