Please login to your online account to display your discounted pricing
Eppendorf™ Multiporator™ Electroporation System
Pulse times in the microsecond (μsec.) range minimize damage to eukaryotic cells
Manufacturer: eppendorf™  940000505
 View more versions of this product
Catalog No. E4308000015
  • / Each
Description & Specifications
Specifications
Includes RS232 interface
Pulse Form Exponentially diminishing
For Use With (Application) Mammalian and plant cell
Certifications/Compliance CE, UL, CAS
Voltage 20/1200V