Eppendorf™ Multiporator™ Electroporation System
Pulse times in the microsecond (μsec.) range minimize damage to eukaryotic cells
$9,547.31 - $9,547.31
Includes: RS-232 interface
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
E4308000015 Eppendorf™
940000505
Each for $9,547.31
Specifications
Description
Hypo-osmolar buffer system increases transfection efficiency; enables electro-deformation of the cells for membrane permeation at specific locations. Soft Pulse™ Technology produces exceptionally high survival rates and greater transfection efficiency.
  • Patented electronics1 ensure accurate performance of pulse parameters, independent of the sample resistance
  • Microprocessor-controlled discharge provides high reproducibility
  • Built-in keypad and display screen allow the user to easily set and monitor operating parameters
Order Info
Eppendorf electroporation cuvets are available separately