Please login to your online account to display your discounted pricing

Eppendorf™ MTP 96 Microplates

Unique clarity for your laboratory

$507.10 - $632.74

Specifications

Autoclavable Yes
Height (Metric) 1.44cm
Material Polypropylene
Type 96-Well Assay Microplate
Height (English) 0.57 in.
View More Specs
Products
Catalog Number Mfr. No. Volume (Metric) Working Color Sterile Volume (Metric) Well Price Quantity    

E951040005

 
eppendorf™
951040005
50 to 350μL Clear No 400μL Each for $507.10

E951040021

 
eppendorf™
951040021
50 to 350μL Clear Yes 400μL Each for $531.67

E951040081

 
eppendorf™
951040081
20 to 320μL Clear Yes 360μL Each for $531.67

E951040308

 
eppendorf™
951040308
20 to 320μL White No 350μL Each for $632.74
Description & Specifications

Specifications

Autoclavable Yes
Height (Metric) 1.44cm
Material Polypropylene
Type 96-Well Assay Microplate
Height (English) 0.57 in.
Width (English) 5.03 in.
Width (Metric) 12.77cm

Ideal for storing substances or applications requiring high resistance to solvens, such as DMSO. The high level of transparency ensures that you always find your samples at a glance.

  • OptiTrack labeling: 30% faster well identification and fewer pipetting errors due to colored border with high-contrast alphanumeric labeling
  • RecoverMax well design: Optimized well geometry for maximum sample recovery and excellent mixing performance
  • G-Safe™ exceptional centrifugation stability of up to 6,000 × g (5,000 × g for sterile plates)
  • Popypropylene provides high resistance to chemicals, mechanical stress and temperature extremes
  • Smooth contours reduce capillary effects in well corners (wicking), minimizing the risk of cross-contamination

  • Manufactured without the use of slip agents, plasticizers and biocides
  • Perfect for automation due to rigid design and dimensional accuracy
  • Minimal residual volume and high well-to-well homogeneity
  • Raised well rims and even surface for reliable sealing
  • No well nesting for easy stacking
  • Good sample visibility

Compliant to SBS specifications

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.