missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eppendorf™ MTP 96 Microplates
Unique clarity for your laboratory
Supplier: Eppendorf™ 951040005
Description
Ideal for storing substances or applications requiring high resistance to solvens, such as DMSO. The high level of transparency ensures that you always find your samples at a glance.
- OptiTrack labeling: 30% faster well identification and fewer pipetting errors due to colored border with high-contrast alphanumeric labeling
- RecoverMax well design: Optimized well geometry for maximum sample recovery and excellent mixing performance
- G-Safe™ exceptional centrifugation stability of up to 6,000 × g (5,000 × g for sterile plates)
- Popypropylene provides high resistance to chemicals, mechanical stress and temperature extremes
- Smooth contours reduce capillary effects in well corners (wicking), minimizing the risk of cross-contamination
- Manufactured without the use of slip agents, plasticizers and biocides
- Perfect for automation due to rigid design and dimensional accuracy
- Minimal residual volume and high well-to-well homogeneity
- Raised well rims and even surface for reliable sealing
- No well nesting for easy stacking
- Good sample visibility
Compliance
Compliant to SBS specifications
Specifications
Standard | |
Autoclavable | |
Polypropylene | |
96 | |
Non-sterile | |
Storing substances or for applications requiring high resistance to solvents, colorimetric assays | |
12.77 cm |
8 x 12 | |
Clear | |
400 μL | |
50 to 350 μL | |
Flat Bottom (F-well) | |
80 Plates | |
1.44 cm |