Eppendorf™ Microcentrifuge Tube Racks
Stackable polycarbonate racks
$63.39 - $63.39
Specifications
Material Polycarbonate
Positions 24
Type Rack, Microcentrifuge Tube
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Autoclavable Holds Price Quantity  
Catalog Number Mfr. No. Autoclavable Holds Price Quantity  
05-405-4 Eppendorf™
022364243
No 0.5mL Tubes Each for $63.39
Specifications
Description
  • Can be interlocked securely front-to-back or side-to-side
  • Numbered tube positions space and elevate tubes properly, making sample identification easy
  • Fully autoclavable racks are UV and chemical resistant, and can be stored at temperatures to -80°C (-176°F)
Capacity: Each rack holds 24 tubes, sizes 0.5mL or 1.5 to 2.0mL.
Order Info
Dimensions (L x W x H): 10.375 × 2.625 × 2.375 in. (26.5 × 6.5 × 6.0cm)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Polycarbonate
24
Rack, Microcentrifuge Tube