Eppendorf™ Membrane Filters for Easypet™ 3 Electronic Pipet Controller
Filters for the Easypet 3 Electronic Pipet Controller
Manufacturer:  Eppendorf™ 4421601009
 View more versions of this product
Catalog No. 12-654-101
Specifications
Description
Filter, Membrane; Eppendorf; Sterile; For use with Easypet 3 Electronic Pipet Controller; 0.45um; Pack of 5
  • Pack of five
  • For Easypet 3
  • Sterile
  • PTFE
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Easypet™ 3 electronic pipet controller
PTFE
0.45μm
Yes