missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ Mastercycler™ pro PCR System

Proprietary vapo.protect™ technology reduces evaporation to a minimum, improving reproducibility and specificity

$4,052.84 - $4,052.84

Specifications

For Use With (Application) PCR applications
Warranty 2 Years
Electrical Requirements 115/230V 50/60Hz
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Product Type Price Quantity  
Catalog Number Mfr. No. Includes Product Type Price Quantity  
E950040035 Eppendorf™
950040035
Mastercycler pro Gradient 384 w/Control Panel N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
E950030010 Eppendorf™
950030010
SteadySlope™, heated Lid Mastercycler pro N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
E950030020 Eppendorf™
950030020
SteadySlope™, heated Lid Mastercycler pro S N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
E950030030 Eppendorf™
950030030
SteadySlope™, heated Lid Mastercycler pro Gradient 384 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
E950030050 Eppendorf™
950030050
Connection cable Mastercycler pro Control Panel Each for $4,052.84
E950040015 Eppendorf™
950040015
SteadySlope™, control panel, heated Lid Mastercycler pro Gradient w/Control Panel N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
E950040025 Eppendorf™
950040025
SteadySlope™, control panel, heated Lid Mastercycler pro Gradient S w/Control Panel N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Key Features

  • Gradient function simplifies protocol optimization
  • Cyclers can be used in three configurations: stand-alone system, mini-satellite systems, or a PC-controlled network
  • Heated lid with integrated Thermal Sample Protection (TSP) technology
  • Intuitive graphic programming
  • SteadySlope™ technology ensures constant heating and cooling rates

  • vapo.protect technology to reduce evaporation
  • Extremely fast heating/cooling rates
  • Gradient blocks with SteadySlope technology; intuitive graphic programming
  • Heated lid with integrated Thermal Sample Protection (TSP) technology
  • Use as stand-alone devices, mini satellite systems or a PC-controlled network
  • Two-year warranty
Mastercycler pro Gradient
  • Freely programmable temperature gradient over 12 rows
  • Gradient range of up to 20°C
  • Robust, specially coated aluminum block
Mastercycler pro Gradient S
  • Highest temperature control speed due to silver block: up to 8°C/sec. with impulse feature
  • Freely programmable temperature gradient over 12 rows
  • Gradient range of up to 24°C
  • Optimized tube fit for fastest heat transition
Mastercycler pro 384
  • Freely programmable temperature gradient over 24 rows
  • Gradient range of up to 20°C

Warranty

Two years

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

PCR applications
2 Years
115/230V 50/60Hz