Eppendorf™ Mastercycler™ pro PCR System
Proprietary vapo.protect™ technology reduces evaporation to a minimum, improving reproducibility and specificity
Manufacturer:  Eppendorf™ 950030030
 View more versions of this product
Catalog No. E950030030
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications
Specifications
≤±0.4°C at 95°C
1 PCR plate
1° to 20°C
SteadySlope™, heated Lid
±0.2°C
115/230V
41.5cm
2 Years
37cm
14.56 in.
50/60Hz
16.33 in.
4° to 99°C
Mastercycler pro Gradient 384
Aluminum
1 x Centronics™, 1 x RS232, control panel, one each of CAN_in/CAN_out
26cm
PCR applications
40.8 lb.
950w
10.23 in.
115/230V 50/60Hz
4°C/sec. heating; 3°C/sec. cooling
18.51kg