Eppendorf™ Mastercycler™ Nexus Thermal Cycler
Offers reliability and ease of use along with low energy consumption
Manufacturer:  Eppendorf™ 6346000021
 View more versions of this product
Catalog No. E6346000021
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Easy expansion to three units with a small footprint fits perfectly on any lab bench. The flexlid accommodates all types of consumables, from low volume all the way up to 0.5mL PCR tubes.
  • Flexlid concept allows use of all types of consumables with automatic height adjustment of the lid
  • Intuitive graphic programming
  • Small footprint
  • Universal block for ultimate flexibility
  • Connect up to two Mastercycler Nexus Eco to your existing unit
  • Available with 12-column SteadySlope gradient
  • Email notification to let you know the status of a run
  • Protocol library for ease of use
  • Booking schedule that acts as screensaver
Note:
The eco unit (Cat. No. E6330000021) is not a standalone unit.  The eco unit must be connected to either the nexus gradient or nexus nongradient unit.
Specifications
Specifications
120V
96 x 0.2mL, 71 x 0.5mL
Aluminum
4° to 99°C
≤±0.4°C at 95°C
Nexus SX1; Silver block
50/60Hz
3°C/sec. heating; 2°C/sec. cooling
±0.2°C
120V 50/60Hz
Documents