Eppendorf™ Mastercycler™ Nexus Thermal Cycler
Offers reliability and ease of use along with low energy consumption
Supplier: Eppendorf™ 6334000026
Description
Easy expansion to three units with a small footprint fits perfectly on any lab bench. The flexlid accommodates all types of consumables, from low volume all the way up to 0.5mL PCR tubes.
- Flexlid concept allows use of all types of consumables with automatic height adjustment of the lid
- Intuitive graphic programming
- Small footprint
- Universal block for ultimate flexibility
- Connect up to two Mastercycler Nexus Eco to your existing unit
- Available with 12-column SteadySlope gradient
- Email notification to let you know the status of a run
- Protocol library for ease of use
- Booking schedule that acts as screensaver
Note: The eco unit (Cat. No. E6330000021) is not a standalone unit. The eco unit must be connected to either the nexus gradient or nexus nongradient unit.
Specifications
96 x 0.2 mL, 71 x 0.5 mL | |
4 to 99°C | |
120 V | |
Nexus Gradient eco | |
3°C/s. Heating; 2°C/s. Cooling |
±0.2°C | |
≤±0.4°C at 95°C | |
120 V, 50/60 Hz | |
50 to 60 Hz | |
Aluminum |