missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ LoBind PCR Plates

Improve recovery of nucleic acids by reducing their absorption to the tube wall. A combination of special manufacturing technologies and selected polypropylene batches ensures nearly 100% recovery of DNA/RNA molecules-without surface coating to eliminate the risk of sample contamination.

Manufacturer:  Eppendorf™ 951040589

 View more versions of this product

Catalog No. E951040589



Description

Description

  • LoBind material guarantees maximum sample recovery for improved assay results
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: of human DNA, DNAase, RNase and PCR inhibitors
  • Available in tube, microplate, and deepwell plate formats for easy-up scaling
  • Unique OptiTrack™ matrix: 30 % faster sample identification and fewer pipetting errors
  • RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties
  • Raised well rims and a smooth surface guarantee reliable sealing in plates
Specifications

Specifications

White
Yes
0.57 in.
30000xG
Yes
5.03 in.
12.77cm
Non-sterile
8.55cm
Protein, peptide, antibody or virus samples
Polypropylene
384
140μL
1.44cm
384 Deep-Well Microplate
Documents