Please login to your online account to display your discounted pricing

Eppendorf™ LoBind PCR Plates

Manufacturer: eppendorf™  925000105

 View all options for this product

Catalog No. E925000105


Description & Specifications


Color White
For Use With (Application) DNA or RNA applications in forensic analysis, microarrays and next generation sequencing
Material Polypropylene
No. of Wells 384
Surface Coating Siliconization
Type 384 Deep-Well Microplate
Volume (Metric) Well 120μL