Please login to your online account to display your discounted pricing

Eppendorf™ LoBind PCR Plates


Manufacturer: eppendorf™  925000103

 View more versions of this product

Catalog No. E925000103

  • / Pack
 

Description & Specifications

Specifications

Color White
For Use With (Application) DNA or RNA applications in forensic analysis, microarrays and next generation sequencing
Material Polypropylene
No. of Wells 96
Surface Coating Siliconization
Type 96 Deep-Well Microplate
Volume (Metric) Well 1000μL