Please login to your online account to display your discounted pricing

Eppendorf™ LoBind PCR Plates


Manufacturer: eppendorf™  925000102

 View more versions of this product

Catalog No. E925000102

  • / Pack
 

Description & Specifications

Specifications

Type 384 Deep-Well Microplate
No. of Wells 384
Volume (Metric) Well 120μL
For Use With (Application) Protein, peptide, antibody or virus samples
Color White
Material Polypropylene
Surface Coating Siliconization
Material Polypropylene
Volume (Metric) Well 120μL